Learn More
Medchemexpress LLC HY-115688A 5mg Medchemexpress, (S)-TXNIP-IN-1 CAS:1212421-96-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY115688A5MG
Encompass_Preferred
Medchemexpress, HY-115688A 5mg (S)-TXNIP-IN-1 CAS:1212421-96-9 (S)-TXNIP-IN-1 is the less active S-enantiomer of TXNIP-IN-1 (HY-115688). TXNIP-IN-1 is a TXNIP-TRX complex inhibitor which can be used in the research of TXNIP-TRX complex associated metabolic disorder (diabetes), cardiovascular disease, or inflammatory disease Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0885
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.