Learn More
Medchemexpress LLC S-ZINC-3573 25mg | 2095596-11-3 | 307.39 | C18H21N5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11568225MG
Encompass_Preferred
(S)-ZINC-3573 is the S-enantiomer of ZINC-3573 supplied for use as an inactive enantiomeric control in pharmacological and cellular studies of primate-selective Mas-related G-protein-coupled receptors (MRGPRs), including MRGPRX2. It is provided as a high-purity solid suitable for biochemical and cellular assays.
  • High purity of 99.89%.
  • Molecular formula C18H21N5 and molecular weight 307.39.
  • Solid, light yellow to yellow appearance.
  • Available in small research quantities such as 25 mg and as a 10 mM solution in DMSO.
  • Intended as an inactive enantiomer control for GPCR/MRGPR research.
Catalog No. 50-003-07924