Learn More
Medchemexpress LLC S-ZINC-3573 50mg | 2095596-11-3 | 307.39 | C18H21N5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11568250MG
Encompass_Preferred
S-enantiomer of ZINC-3573 used as an inactive negative control in studies of the primate-specific MRGPRX2 receptor. Provided as a high-purity solid for research use, available in small lab-scale quantities and in DMSO solution formats. Follow the supplier-recommended storage and handling guidelines.
Catalog No. 50-003-07925