Learn More
Thermo Scientific™ S1 Pipet Filler Accessories
GREENER_CHOICE
Catalog No. 14387043
Change view
Click to view available options
Type:
Nosepiece
Pipette Filter, Non-sterile
Pipette Filter, Sterile
Silicone Pipet Gripper
Table Mount Stand
Universal Power Supply
Wall Mount Stand
Wing Stand
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-387-043 Universal Power Supply
14-386-77 Pipette Filter, Non-sterile
14-387-038 Pipette Filter, Sterile
14-387-039 Nosepiece
14-387-040 Silicone Pipet Gripper
14-387-041 Wall Mount Stand
14-387-042 Table Mount Stand
14-388-500 Wing Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 14-387-043 Supplier Thermo Scientific™ Supplier No. 9068
Add to Cart
Add to Cart
This item is not returnable. View return policy
Incorporate Thermo Scientific™ S1 Pipet Filler Accessories with S1 Pipet Filler.
Complete your S1 Pipet Filler with these accessories and prolong its use.
Features:
  • Wall-mounted holder and table stand offer compact storage between applications. Recharge the pipet filler in both stands by plugging in the charger. Store pipet filler when working in laminar flow cabinets to save space.
  • Wing stand allows user to set the pipet filler down without removing or emptying the serological pipet.
  • Easy to change filter prevents aerosol or liquid cross contamination of the pipet together with autoclavable silicone adapter.
  • 1mL Pipet Support ensures that 1 mL pipets will hold as securely as larger sizes.
Note:
Wing stand not available for use or sale within the United States or for importation into the United States.
TRUSTED_SUSTAINABILITY
Specifications
For Use With S1 Pipette Filler
Type Universal Power Supply