Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100 Recombinant Rabbit Monoclonal Antibody (JM51-31)
GREENER_CHOICE

Catalog No. PIMA532625 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532625 Supplier Invitrogen™ Supplier No. MA532625
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JM51-31
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene S100A1
Gene Accession No. P04271, P23297, P50114, P56565
Gene Alias AI266795; AI850290; Bpb; hypothetical protein LOC525716; NEF; Protein S100-A1; protein S100-B; S 100; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium binding protein B; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 protein, alpha polypeptide; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100a; S100A1; S100alpha; S100-alpha; S100B; S100-B; S100beta; S100P
Gene Symbols S100A1, S100B
Host Species Rabbit
Immunogen Recombinant full length protein of mouse S100 aa 1-94.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20193, 20203, 6271, 6285
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.