Learn More
Description
Specifications
Specifications
| Assay Range | 31.25 pg/mL |
| Assay Sensitivity | 31.25-2000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse S100A1. |
| Target Species | Mouse |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
