Learn More
MBL International Corporation CircuLex S100A13 ELISA Kit
SDP
Supplier:  MBL International Corporation CY8057
Encompass_Preferred
Application:ELISA; Size:96 Assays; Target:S100A13; The  CircuLex S100A13 ELISA Kit is used for the quantitative measurement of human S100A13 in tissue culture medium and other biological media. It can be used for 100 assays.CircuLex S100A13 ELISA Kit.
Catalog No. 50-455-98
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.