Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ PB-22 3-carboxyindole metabolite Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco S1021ML
Description
This Certified Spiking Solution™ is applicable for use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology and forensic analysis. PB-22 3-carboxyindole metabolite is a major metabolite of PB-22 found in serum and urine. PB-22 is an analog of the potent synthetic cannabinoid JWH 018.Specifications
75-05-8, 727421-73-0 | |
MFCD05859192 | |
100 μg/mL in acetonitrile | |
Ampule | |
2°C (35.6°F) - closed cup |
C14H17NO2 | |
UN 1648 3 / PGII | |
231.29 | |
1 mL |
Safety and Handling
H332-H319
H312
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?
Provide Content Correction