Learn More
Thermo Scientific S120-AT3 FIXED ANGLE ROTOR
Supplier:  Thermo Scientific 45584
Encompass_Preferred
Rotor, Fixed-Angle; Thermo Scientific; S120-AT3; For Thermo Scientific Sorvall MTX 150 Series Micro-Ultracentrifuge, MX Series Floor Model Micro-Ultracentrifuge; Capacity: 14 x 0.5mL (0.08L); Titanium; Max. speed/RCF: 120,000rpm/649,826 x g
Catalog No. 50-870-323
Only null left
Add to Cart
Request a Quote