Learn More
CPC Scientific H-Lys-Glu-Ala-Lys-Glu-Lys-Arg-Gln-Glu-Gln-Ile-Ala-Lys-Arg-Arg-Arg-Leu-Ser-Ser-Leu-Arg-Ala-Ser-Thr-Ser-Lys-Ser-Gly-Gly-Ser-Gln-Lys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PKCP047A
Encompass
Peptide that measures the activity of kinases that phosphorylate ribosomal protein S6. It can also be used as a substrate.ONE-LETTER SEQUENCE: KEAKEKRQEQIAKRRRLSSLRASTSKSGGSQKMOLECULAR FORMULA: C149H270N56O49MOLECULAR WEIGHT:3630.2STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0199
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.