Learn More
Selleck Chemical LLC Dubermatinib(TP-0903) 5mg 1341200-45-0
SDP
Supplier:  Selleck Chemical LLC S78465MG
Encompass_Preferred
TP-0903 is a potent and selective AXL Inhibitor with IC50 of 27 nM. TP-0903 is highly effective in inducing apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1151703
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.