Learn More
Coast SA300 High-Vis Lighted Safety Armband
Catalog No. 19012107
Change view
Click to view available options
Size:
One Size
Catalog No. Size
19-012-107 One Size
1 options
Catalog No. 19-012-107 Supplier Coast Supplier No. 30399
Increase safety in dark and chaotic environments with the SA300 Armband which provides up to 10 hours of light on a single charge, and two modes (solid and flashing) of highly visible orange light.
  • Versatile, rechargeable safety light can be worn on the body during work or play
  • Provides highly visible orange safety light in two modes, solid and flashing
  • On a single charge, the light can run for up to 10 hours on solid mode and 12 hours on flashing mode
  • Constructed of water-resistant material to ensure a long life
  • Designed with an adjustable strap and hook system for no-pinch comfort
  • Armband is constructed with flexible straps so that it can be placed right where it is needed - on the upper arm, wrist, around the knee, or ankle
  • Easily recharge the armband with the included charging cable
  • LED light can be removed when the band needs washing
Specifications
Size One Size
Width (English) 2.13 in.
Width (Metric) 5.39 cm
Color Hi-vis Orange
Product Type Armband