Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sabouraud Dextrose Agar

Catalog No. R112562
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
20 x 150mm Pour Tube (G-PT)
20mm x 150mm Tube - Slant
BB - 200mL
BB - 500mL
BRB - 100mL
Monoplate Deep Pour
Manufacturing Origin:
United States
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Manufacturing Origin
R112562 10/Pk. BRB - 100mL United States
R01766 100/Pk. Monoplate Deep Pour United States
R01768 10/Pk. Monoplate Deep Pour United States
R09746 100/Pk. 20mm x 150mm Tube - Slant United States
R112551 10/Pk. BB - 500mL United States
R09744 100/Pk. 20 x 150mm Pour Tube (G-PT) United States
R09747 20/Pk. 20mm x 150mm Tube - Slant United States
R112550 10/Pk. BB - 200mL United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. R112562 Supplier Thermo Scientific™ Supplier No. R112562
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, on a ready-to-use prepared media plate.

Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, using acidic pH Thermo Scientific™ Sabouraud Glucose Agar.

Fungi maintain their typical cultural appearance and may thus be readily identified according to the standard macroscopic characteristics. Infections caused by Candida spp. are diverse, ranging from superficial and local to deep-seated and disseminated. Candida albicans and Candida glabrata comprise 70-80% of yeast isolates recovered from patients with invasive disease. This culture media, also available in powder form, has applications in environmental monitoring and sterility testing, enabling corrective measures to be implemented rapidly if unacceptable levels of microbial loading indicate lapses in process control.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, COMPF, USP
Color Amber
Content And Storage 2 to 25°C
Format BRB - 100mL
Manufacturing Origin United States
Product Type Agar Plate
Fill Volume 100 mL
Packaging Type 10 x 100mL
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.