Learn More
Bioassay Systems Saccharide Removal Kit, For rapid removal of interfering saccharides (e.g. glucose, sucrose) from samples such as culture media, wines, beers, fermented teas.. Treated samples can be directly used for ethanol determination with DIET-500 assay kit.
SDP
Catalog No. 5000144366 Shop All Bioassay Systems Products
Encompass_Preferred
SACCHARIDES such as glucose and sucrose are known to interfere with the QuantiChrom™ Ethanol Assay (catalog# DIET-500). BioAssay Systems has developed a rapid procedure for the complete removal of saccharides by co-precipitation with alkaline cupric and calcium ions. The treatment takes as little as 5 min and removes >99.9% saccharide from a sample.
Catalog No. 50-001-44366 Supplier Bioassay Systems Supplier No. DSRK2500
SACCHARIDES such as glucose and sucrose are known to interfere with the QuantiChrom™ Ethanol Assay (catalog# DIET-500). BioAssay Systems has developed a rapid procedure for the complete removal of saccharides by co-precipitation with alkaline cupric and calcium ions. The treatment takes as little as 5 min and removes >99.9% saccharide from a sample.