Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharin Calcium, Crystal, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18606113 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 lb.
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-113 100 lb. Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-606-113 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CA167100LBBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

CA167, 6381-91-5, C14H8CaN2O6S2•31/2H2O

Spectrum™ Chemical Saccharin Calcium, Crystal, USP is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6381-91-5
Molecular Formula C14H10CaN2O7S2
Molecular Weight (g/mol) 422.44
InChI Key XOCXGVIOTKFUSU-UHFFFAOYSA-L
IUPAC Name calcium bis(1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate) hydrate
SMILES O.[Ca++].[O-]S1(=O)=NC(=O)C2=CC=CC=C12.[O-]S1(=O)=NC(=O)C2=CC=CC=C12

Specifications

CAS Max % 1
Quantity 100 lb.
Percent Purity 99 to 101%
Grade USP
Packaging Fiber Drum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.