Learn More
ATCC Saccharomyces cerevisiae (strain 641a) - ATCC 24860, freeze-dried culture
Supplier: ATCC 24860
Saccharomyces cerevisiae (strain 641a) is a wild-type diploid yeast supplied as a viable, freeze-dried culture for laboratory propagation and fermentation studies. It is suitable for research in carbohydrate metabolism, ethanol production, and xylulose fermentation.
- Supplied as a freeze-dried viable culture for propagation.
- Strain: 641a (VTT C-79096).
- Recommended storage: 2°C to 8°C.
- Suitable growth media: YM agar or broth, Emmons' modification of Sabouraud's agar/broth, YEPD.
- Common applications: xylulose and glucose fermentation studies, transaldolase production, ethanol-tolerance research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.