Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Saccharomyces cerevisiae ZIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117585
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117585 Supplier Invitrogen™ Supplier No. PA5117585
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

ZIP1, also known as ZIRTL (zinc-iron regulated transporter-like), is the first mammalian member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP1 expression is markedly downregulated in a number of cancerous tissues and is thought to function as a tumor suppressor gene in prostate cancer. More recent studies have shown that overexpression of ZIP1 and concomitant increased levels of intracellular zinc in PC-3 cells cause a significant inhibition of NF-kappa-B, leading to down-regulation of the antiapoptotic protein Bcl-2, Bcl-XL, and the apoptosis inhibitor XIAP, reducing the malignant potential of prostate cancer cells in vitro and in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Saccharomyces cerevisiae ZIP1
Applications Western Blot
Classification Polyclonal
Concentration 2.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZIP1
Gene Accession No. P31111
Gene Alias CGI-08; CGI-71; IRT1; mZIP1; SLC39A1; solute carrier family 39 (zinc transporter), member 1; solute carrier family 39 (zinc transporter), member 3; solute carrier family 39 member 1; Zinc transporter ZIP1; Zinc-iron-regulated transporter-like; Zip1; ZIP-1; ZIRTL; Zrt- and Irt-like protein 1; Zrt/Irt-like protein 1
Gene Symbols ZIP1
Host Species Rabbit
Immunogen Recombinant Saccharomyces cerevisiae Synaptonemal complex protein ZIP1 protein (1-227aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 851879
Target Species Yeast
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.