Learn More
Apexbio Technology LLC SacI 2000U
SDP
Supplier:  Apexbio Technology LLC K30432000
Encompass_Preferred
SacI is a recombinant restriction endonuclease targeting double-stranded DNA at the palindromic sequence 5 -GAGCT C-3 It is designed to cleave DNA generating cohesive ends conducive to molecular biology workflows SacI exerts its biological activity primarily through sequence-specific endonucleolytic cleavage Based on these properties SacI holds research potential in applications such as restriction digestion analyses plasmid screening insert verification and genomic DNA manipulation
Catalog No. 50-000-97231
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.