Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SAE Administrator Console Software v2.1

Catalog No. A53717
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 license
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53717 1 license
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A53717 Supplier Thermo Scientific™ Supplier No. A53717

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or visit www.fishersci.com/quantstudio for more information and to contact a Life Science Specialist.

SAE Administrator Console Software v2.1 provides security, audit, and electronic signature (SAE) for the Applied Biosystems SeqStudio Flex Series genetic analyzers. The software supports compliance with guidelines on electronic records security, and prevents unauthorized access to the instrument.

SAE Administrator Console Software v2.1 provides security, audit, and electronic signature (SAE) for the Applied Biosystems SeqStudio Flex Series genetic analyzers. The software supports compliance with guidelines on electronic records security and prevents unauthorized access to the instrument.

System Security

  • Controls user access to the software
  • A default Administrator user account is provided
  • Additional user accounts and permissions can be user-defined

Auditing

  • Tracks actions performed by users, and changes to the SAE module settings
  • The software automatically audits some actions silently
  • Users can select other items for auditing and specify the audit mode
  • The auditing function provides reports for audited SAE module changes and actions

Electronic signature (e-signature)

  • Determines if users are required to provide a user name and password when performing certain functions
  • Configure an e-signature so that a user can print a report or start a run only if the associated data are signed
  • Also, configure each e-signature event to require multiple signatures and to require users with specific permissions to sign
TRUSTED_SUSTAINABILITY

Specifications

Product Line SeqStudio Flex
Software Type Sequencing and Fragment Analysis Software
Content And Storage The software is provided in the form of a USB stick.
For Use With (Equipment) Sequencing Instruments (Capillary Electrophoresis-Based)
Product Type SAE Administrator Console Software v2.1
Quantity 1 license
Shipping Condition Room Temperature
Where can I download the Release Notes for SAE Administrator Console Software v2.1?

Please download this PDF (http://downloads.thermofisher.com/SAE_AdminConsole_v2.1/USB/100108548_SeqStudioFlex_SAE_Admin_Console_Software_Version_2_1_0_Release_Notes.pdf).

I created a user profile in SAE Administrator Console Software v2.0. Can I install it on SAE Administrator Console Software v2.1?

Yes, application profiles created with earlier versions of the SAE software can be used with later versions of the SAE software. However, profiles created with newer SAE servers are not supported by earlier versions.

What applications are compatible with the SAE Administrator Console Software v2.1?

The following applications are compatible with the SAE Administrator Console Software v2.1:

- SeqStudio Flex Series Instrument Software v1.0 and above
- Design and Analysis Software v2.5.0 and above
- QuantStudio 6/7 Pro Version 1.4.0 Instrument Software and above

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.