Learn More
Mopec™ Safe Seal System Pre-cut Body Bags Starter Pack

The Safe Seal System of these Pre-cut Body Bags is a military tested, heavy duty, hermetically sealable Bio-Safety Level 4 (BSL-4) Containment body bag system. It is easy to use, as the body is placed in the flexible material, then heat-sealed with the Safe Seal Heat Sealer to provide complete and secure containment of infectious human or animal remains.
Supplier: Mopec™ BE160
Description
The Safe Seal System was designed to help keep workers and survivors safe after a mass fatality event or any high-risk biohazard exposure. The Starter Pack contains the following:
- 25 Pre-cut, 48 x 96 in. Bags
- 1 Heat Sealer
- 2 Alignment Clips
Specifications
>200 lb. | |
Gray | |
8 mil | |
96 in. | |
48 in. | |
Body Bag |
Safe Seal | |
Plastic | |
96 x 48 in. | |
243.84 cm | |
121.92 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.