Learn More
Mopec™ Safe Seal System Replacement Pre-cut Body Bags

The Safe Seal System of these Replacement Pre-cut Bags is a military tested, heavy duty, hermetically sealable Bio-Safety Level 4 (BSL-4) Containment body bag system. It is easy to use, as the body is placed in the flexible material, then heat-sealed with the Safe Seal Heat Sealer to provide complete and secure containment of infectious human or animal remains.
Supplier: Mopec™ BE161
Description
The Safe Seal System was designed to help keep workers and survivors safe after a mass fatality event or any high-risk biohazard exposure.
- Contains 25 Bags/Cs.
Specifications
>200 lb. | |
Gray | |
243.84 cm | |
8 mil | |
96 in. | |
48 in. | |
Body Bag |
Safe Seal | |
96 in. | |
Plastic | |
96 x 48 in. | |
243.84 cm | |
121.92 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.