Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Safe-Sense™ Blue Nitrile Powder Free Chemo Rated Exam Gloves
SDP

Catalog No. 18048002 Shop All Medisca Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-048-002 Large
18-048-000 Small
18-048-001 Medium
18-048-003 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-048-002 Supplier Medisca Supplier No. 574402
Add to Cart
Add to Cart
This item is not returnable. View return policy

Medisca Safe-Sense™ Powder Free Chemo Rated Exam Gloves provide maximum protection for non-sterile hazardous compounding, administration, disposal and other related chemotherapy activities. They are resistant to puncture, abrasion and snag.

  • Medical examination gloves
  • Improves user and product protection for superior durability
  • Polyurethane coating for easy donning and removal
  • Beaded cuff to reduce glove roll down and ripping
  • Textured fingertips to provide grip and sensitivity
  • Produced with distilled water for a silky feeling
  • Disposable
  • Meets EN455 standard and ASTM D6319
  • Meets ASTM D6978 resistant to permeation by chemotherapy drugs
  • Manufactured in an ISO 13485 certified facility
  • Meets FDA regulations for food handling applications

Specifications

Certifications/Compliance ASTM D6978-05
Certifications/Compliance (Europe) EN455
Chemotherapeutic Rated Yes
Applications Healthcare, Laboratory, Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Large
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
FDA 510 (k) Yes
Quantity 100 Gloves/Box, 10 Boxes/Cs.
Length (English) 9 in.
Length (Metric) 22.86 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.