Learn More
RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: Serum
Description
Includes
Pre-assembled capillary blood collection device with a capillary tube, attached pierceable cap, and a microtube
Blood samples are drawn into the SAFE-T-FILL™ Plastic Micro Capillary Blood Collection Tubes through capillary action, reducing hemolysis, tissue contamination, and re-draws. No scooping or milking required. Coated with Sodium Fluoride for glucose determinations.
- 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
- Blood samples are drawn into the microtube through capillary action, reducing hemolysis, tissue contamination, and re-draws. No "scooping" or "milking" required
- Capillary straw and microtube are coated with anticoagulant Lithium Heparin (Green cap) to prevent clotting
- Attached cap is color-coded to designate the type of additive that is in both the capillary tube and the microtube
- Lot Number, Expiration Date, and fill lines are printed directly on the microtube
- A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
- For plasma determinations in chemistry
- Optional gel separator serves as a barrier between blood and serum after centrifugation
- Available in a variety of sizes and draw volumes, with or without gel separator
- Optional amber microtube ensures light-sensitive sample integrity providing protection for testing (ex. Bilirubin)
Recommended Storage
Store at room temperature. Keep dry. Keep away from sunlight.
Specifications
Specifications
| Color | Clear |
| Product Type | Chemistry/Serum Tube |
| Additive | Clot Activator |
| Material | Plastic |
| Closure Type | Standard Pierceable Cap |
| Capacity (Metric) | 200 μL |
| Closure Included | Yes |
| Closure Color | Red |
| For Use With (Application) | Intended for chemistry determinations that require serum or clotted blood samples. |
| Description | Serum Gel Capillary Collection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.