Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: Serum

Catalog No. 1491559
Change view
Click to view available options
Capacity (Metric):
200 μL
300 μL
Additive:
Clot Activator
Clot Activator and Gel
Color:
Amber
Clear
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Additive Color
14-915-59 200 μL Clot Activator and Gel Clear
14-915-56 200 μL Clot Activator Clear
14-915-57 300 μL Clot Activator Clear
14-915-60 300 μL Clot Activator and Gel Clear
14-915-63 200 μL Clot Activator and Gel Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 14-915-59 Supplier RAM Scientific Supplier No. 07 7120
This item is not returnable. View return policy

Includes

Pre-assembled capillary blood collection device with a capillary tube, attached pierceable cap, and a microtube

No scooping needed, allowing for high-quality, tissue-free samples

Blood samples are drawn into the SAFE-T-FILL™ Plastic Micro Capillary Blood Collection Tubes through capillary action, reducing hemolysis, tissue contamination, and re-draws. No scooping or milking required. Coated with Sodium Fluoride for glucose determinations.

  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Blood samples are drawn into the microtube through capillary action, reducing hemolysis, tissue contamination, and re-draws. No "scooping" or "milking" required
  • Capillary straw and microtube are coated with anticoagulant Lithium Heparin (Green cap) to prevent clotting
  • Attached cap is color-coded to designate the type of additive that is in both the capillary tube and the microtube
  • Lot Number, Expiration Date, and fill lines are printed directly on the microtube
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • For plasma determinations in chemistry
  • Optional gel separator serves as a barrier between blood and serum after centrifugation
  • Available in a variety of sizes and draw volumes, with or without gel separator
  • Optional amber microtube ensures light-sensitive sample integrity providing protection for testing (ex. Bilirubin)

Recommended Storage

Store at room temperature. Keep dry. Keep away from sunlight.

Specifications

Color Clear
Product Type Chemistry/Serum Tube
Additive Clot Activator and Gel
Material Plastic
Closure Type Standard Pierceable Cap
Capacity (Metric) 200 μL
Closure Included Yes
Closure Color Red
For Use With (Application) Intended for chemistry determinations that require serum or clotted blood samples. Gel separator serves as a barrier between blood and serum after centrifugation.
Description Serum Gel Capillary Collection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.