Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChemDAQ™ SafeCide™ 2.1 Hydrogen Peroxide Monitor System - Handheld
Description
Key features include:
- Continuously displays real-time measurements of vapor concentrations in parts per million (ppm)
- Displays 15 min and 8 h Time Weighted Average (TWA) exposure readings
- Customizable audible and visual alarms, including impending alerts.
- Features visual (green, yellow, red) and audible alerts to warn when unsafe exposure limits are reached.
- Timely and consistent shipment of calibrated sensors through ChemDAQ's exclusive Sensor Exchange Program (SXP™)
Specifications
Specifications
| Components Measured | Hydrogen Peroxide |
| For Use With (Equipment) | ChemDAQ Gas Sensor |
| Type | Portable Gas Detection Monitor |
Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?