Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ SafeCide™ 2.1 Hydrogen Peroxide Monitor System - Tablet

Catalog No. 17987104
Change view
Click to view available options
Components Measured:
Hydrogen Peroxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-104 Hydrogen Peroxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-104 Supplier ChemDAQ™ Supplier No. PSC2.1HMONT
Add to Cart
Add to Cart

The SafeCide™ 2.1 is a tablet based portable monitor designed for real time detection and logging of hydrogen peroxide (H2O2) vapor concentrations up to 20 ppm with 0.1 ppm resolution.

It features dual operation modes:

  • Environmental Sampling Mode for detecting leaks and hot spots
  • Industrial Hygiene Mode for calculating 8 hour and 15 minute time weighted averages (TWA)

Key features include:

  • A tablet display with built-in SafeCide 2.1 software offering easy viewing of real time readings, summary dashboards, and data export to CSV.
  • Bluetooth connectivity, automatic sensor reconnection, low battery and calibration alerts, and support for multiple user credentials or guest mode.
  • Customizable visual (green/yellow/red) and audible alarms to warn when exposure limits are exceeded.
  • E Cell™ sensor modules backed by ChemDAQ's Sensor Exchange Program (SXP™), automatically reminding users when calibration is due.

This rugged handheld tablet kit is ideal for spot-checking facility environments or performing personal monitoring, especially in sterilization and hygiene-critical settings such as healthcare, pharmaceuticals, and food processing. It helps ensure compliance, enhance safety, and simplify reporting.

Specifications

Components Measured Hydrogen Peroxide
For Use With (Equipment) ChemDAQ Gas Sensor
Type Portable Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.