Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChemDAQ™ SafeCide™ 2.1 MultiGas Monitor System - Tablet
Description
The SafeCide™ portable, handheld monitoring system provides real-time measurements in parts per million, providing accurate, reliable readings.
Features include:
- Two operating modes: Sample Mode and Industrial Hygiene Uses ChemDAQ's E-Cell™ Sensor modules, currently available for hydrogen peroxide
- Displays instantaneous readings, 15-min and 8-hour time weighted average (TWA), to ensure a safe working environment and regulatory compliance
- Customizable audible and visual alarms
- Email alerts to notify you of dangerous levels in real-time
- Data can be stored and analyzed on the SafeCide device or exported to a computer for further analysis and backup storage
Specifications
Specifications
| Components Measured | Hydrogen Peroxide, Peracetic Acid |
| For Use With (Equipment) | ChemDAQ Gas Sensor |
| Type | Portable Gas Detection Monitor |
Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?