Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ SafeCide™ 2.1 Peracetic Acid Monitor System - Tablet

Catalog No. 17987108
Change view
Click to view available options
Components Measured:
Hydrogen Peroxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-108 Hydrogen Peroxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-108 Supplier ChemDAQ™ Supplier No. PSC2.1PMONT
Add to Cart
Add to Cart

The SafeCide™ 2.1 MultiGas Monitor is a handheld system designed for real-time detection of peracetic acid.

The SafeCide™ 2.1 Tablet Peracetic Acid (PAA) Monitor is a high-visibility, real-time gas detection system designed to monitor airborne PAA levels in disinfection and sterilization environments. Featuring a large touchscreen tablet interface, this monitor provides enhanced usability, clear data presentation, and expanded functionality for environmental health and safety (EHS) professionals. With continuous monitoring, customizable alarms, and data logging, the SafeCide™ 2.1 Tablet system ensures worker safety, regulatory compliance, and operational efficiency in environments where PAA is used as a sterilant or disinfectant.

Key Features

  • Tablet Interface with Large: The integrated tablet offers an intuitive, high-resolution touchscreen interface that displays real-time PAA concentrations, time-weighted average (TWA), and short-term exposure limit (STEL) data. The larger screen allows for easier viewing of trends, logs, and alerts, ideal for centralized monitoring stations or facility dashboards.
  • Continuous PAA Monitoring: The system provides live, real-time detection of peracetic acid vapors, helping to protect personnel and maintain compliance with exposure limits from OSHA and NIOSH. Instant feedback supports proactive safety decisions during sterilization or cleaning processes.
  • Built-in Exposure Analytics: Track and review exposure data over time, including TWA and STEL readings. The system alerts users when thresholds are exceeded and logs session data for documentation and compliance auditing.
  • Sensor Exchange Program (SXP™) Compatible: ChemDAQ's SXP™ pre-calibrated sensors allow for quick and simple sensor replacement without tools or downtime. Maintain measurement accuracy while minimizing maintenance efforts.
  • Customizable Alarms: Visual alerts on the tablet screen and optional audible notifications alert staff when peracetic acid levels approach or exceed preset safety thresholds. Alarm points can be configured to match your facility's safety protocols.
  • Data Logging and Export: Session data is stored on the tablet and can be exported for recordkeeping, internal reviews, or regulatory inspections. Helps teams analyze patterns and make informed decisions about workflow safety and environmental controls.

Peracetic acid is a powerful sterilizing agent, but its vapor can pose serious health risks if not carefully monitored. The SafeCide™ 2.1 Tablet Monitor offers enhanced visibility, continuous tracking, and data recording in a streamlined, user-friendly system. Whether stationed in a sterile processing area or deployed in a quality assurance office, this tablet-based system delivers confidence, compliance, and control in environments that demand the highest safety standards.

Specifications

Components Measured Hydrogen Peroxide
For Use With (Equipment) ChemDAQ Gas Sensor
Type Portable Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.