Description
- Dual-fit nose and chin pieces adjust to form a tight seal around the face to reduce gapping and ensure a personalized fit.
- Soft non-woven material enhances breathability.
- Smooth stretchy latex-free ultrasonically sealed earloops prevent irritation and allow easy removal with a lower risk of self-contamination.
Specifications
Specifications
| Product Type | Medical/Procedural Face Mask |
| ASTM Level | ASTM Level 1 |
| Color | Blue |
| Fastening Type | Earloops |
| Sterility | Non-sterile |
| Style | Pleated |
| Disposable | Single-use |
| Latex Free | Yes |