Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safetec™ P.A.W.S. Hand Wipes
SDP

Catalog No. 19819585
Encompass_Preferred
Change view
Click to view available options
Quantity:
100/Pk., 10/Cs.
160 Wipes/Canister, 12/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-819-585 160 Wipes/Canister, 12/Cs.
19-819-578 100/Pk., 10/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-819-585 Supplier Safetec™ Supplier No. 34410
This item is not returnable. View return policy

Ideal for hand sanitizing when soap and water are not available

  • Antimicrobial hand wipes with 66.5% ethyl alcohol to kill 99.9% germs within 15 seconds
  • Contain aloe vera to keep your skin feeling soft and smooth all day long
  • Remove debris and soil from from hands and under fingernails
  • Wipe size: 5 × 8 in.
  • Meet OSHA, APIC and CDC hand washing recommendations in the absence of soap and water

Specifications

Product Type P.A.W.S.™ Hand Wipe
Form Wipe
Certifications/Compliance OSHA, APIC, CDC
Composition Ethyl Alcohol 66.5%, Aloe Vera
For Use With (Application) Ideal for hand sanitizing in the absence of soap and water
Quantity 160 Wipes/Canister, 12/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.