Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Safetec Oral Pain Relief

Catalog No. 19090840 Shop All Medique Products
Change view
Click to view available options
Quantity:
144/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-840 144/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-840 Supplier Medique Supplier No. 53117
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Temporary relieves pain caused by toothache, canker sores, gum sores, dental work, cheek bites, denture irritation and other irritation or minor injury of the mouth and gums.

  • Active ingredient Benzocaine

Specifications

Product Type Safetec Oral Pain Relief
Active Ingredients Benzocaine
For Use With (Application) Relieves pain from toothache, canker and gum sores, denture irritation
Format In 0.75 g Packets
Quantity 144/Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.