Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ SafeTech™ Vapor Monitor
GSA_VASDP

Catalog No. p-7227181
Change view
Click to view available options
Components Measured:
Formaldehyde
Xylene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
STLVMBFORM6 Formaldehyde
STLVMBXYLN6 Xylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. STLVMBFORM6 Supplier StatLab™ Supplier No. VMBFORM/6
This item is not returnable. View return policy

Vapor monitoring is important to ensure the safety of employees and comply with OSHA guidelines. StatLab™ makes it easy to monitor vapor levels of chemicals in your laboratory with these badges.

  • Simply send the badges in and access your results via statlab.com
  • Badges include prepaid analysis for 8 hour or 15 minute exposure readings
  • Complies with OSHA standards for vapor exposure in the workplace

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance OSHA
Components Measured Formaldehyde
For Use With (Application) Measuring Vapor Exposure
Quantity 6 Badges
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.