Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ SafeTech Vapor Monitor
SDP

Catalog No. STLVMBFORM
Change view
Click to view available options
Components Measured:
Formaldehyde
Xylene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
STLVMBFORM Formaldehyde
STLVMBXYLN Xylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. STLVMBFORM Supplier StatLab™ Supplier No. VMBFORM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vapor monitoring is important to ensure the safety of employees and comply with OSHA guidelines.

  • StatLab™ makes it easy to monitor vapor levels in your laboratory
  • Simply send the badge in and access your results via our website
  • Includes prepaid analysis for 8-hour or 15-minute exposure readings
  • Complies with OSHA standards for vapor exposure in the workplace

Specifications

Components Measured Formaldehyde
For Use With (Equipment) Laboratory
Type Single Gas Detector
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.