Learn More
Andwin Scientific Safetex™ No-Touch Pap Kits
SDP
Catalog No. 1437241
Encompass_Preferred
Change view
Click to view available options
Product Type:
No-touch 1-slide Papkit
No-touch Pap Smear Test Kit
Includes:
Cotton Swab, Fixative Pouch
Plastic Scraper
Super-frosted Slide, Fixative Spray
Wood Scraper, Fixative Pouch
Wood Scraper, Fixative Spray
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
14-372-41 No-touch Pap Smear Test Kit Plastic Scraper
14-372-42 No-touch Pap Smear Test Kit Wood Scraper, Fixative Spray
14-373-46 No-touch 1-slide Papkit Cotton Swab, Fixative Pouch
14-372-82 No-touch 1-slide Papkit Super-frosted Slide, Fixative Spray
14-372-36 No-touch Pap Smear Test Kit Wood Scraper, Fixative Pouch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 14-372-41 Supplier Andwin Scientific Supplier No. 230170
Add to Cart
Add to Cart
This item is not returnable. View return policy
Product requires no tape or glue to hold slide
  • 'No-Touch' specimen pack provides maximum slide specimen protection
  • Ventilation folds allow wet slides to dry in courier pack
  • Each Kit includes Slide, Scraper, and Brush, and may include additional accessories.
Specifications
Product Type No-touch Pap Smear Test Kit
Includes Plastic Scraper
For Use With (Application) Provides maximum slide specimen protection
Weight (English) 24 lb.
Weight (Metric) 10.8 kg
Quantity 25 Kits/Pk.