Learn More
Caresfield Safety Blood Collection Needle with Integrated Safety Shield and Pre-Attached Holder
SDP
Catalog No. 22306388
Change view
Click to view available options
Color:
Black
Blue
Green
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-306-388 Green
22-306-389 Black
22-306-390 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-306-388 Supplier Caresfield Supplier No. 61628CS
Only null left
Add to Cart
Add to Cart
The Safety Blood Collection Needle comes with an integrated safety shield and a pre-Attached Holder.
  • Hinged safety shield mechanism
  • Large, ergonomic finger grip
  • Double sheaths
  • Pre-attached holder
  • 500 per case
Specifications
Product Type Safety Blood Collection Needle
Material Stainless Steel
Sterility Sterile
Color Green