Learn More
Eisco™ Safety Face Shield
SCI_ED
Catalog No. S27083 Shop All Eisco Products
Change view
Click to view available options
Lens Coating:
Impact-resistant
Catalog No. Lens Coating
S27083 Impact-resistant
1 options
This item is not returnable. View return policy
Catalog No. S27083 Supplier Eisco™ Supplier No. CH0620
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fits securely to deep molded brow guard with adjustable crown strap for protection against chemical splashes and impact from flying objects
  • Safety Face Shield
  • For use in any home, school, or professional laboratory
  • Fits securely to deep molded brow guard with adjustable crown strap
  • Protects against chemical splashes and impact from flying objects
  • Steel bound plastic visor
  • Dimensions (L x W): 7 x 8 in.
  • Non-flammable
Specifications
Dimensions (L x W) 7 x 8 in.
Lens Coating Impact-resistant
Lens Tint None
Product Type Visor
For Use With (Application) Protection Against Chemical Splashes and Impacts From Flying Objects
Material Steel Mesh, Nylon
Disposable Limited-use
Lens Material Plastic
Frame Material Plastic
UV Protection None