Learn More
Description
- Cork-lined jaws
- Made from stainless-steel
Specifications
Specifications
| Product Type | Tongs |
| Surface Coating | Cork-lined Jaws |
| Length (Metric) | 30 cm |
| Description | Safety flask tongs |
| Material | Stainless Steel |
| For Use With (Application) | Fits flasks of any shape from 100 to 2000mL |
| Includes | 1 set of tongs |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
