Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Safety Flip Chart
SCI_ED

Catalog No. S01469 Shop All NewPath Learning Products
Change view
Click to view available options
Product Type:
All About Animals Flip Chart Set
All About Me Flip Chart Set
All About Plants Flip Chart Set
Curriculum Mastery Flip Chart Set
Exploring Matter Flip Chart Set
Math Skills Flip Chart Set
Our Earth Flip Chart Set
Pushing, Moving and Pulling Flip Chart Set
Scient Readiness Flip Chart Set
Weather and Sky Flip Chart Set
Grade:
6 to 8
7
8
Pre-K to 1
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Grade
S01469 Curriculum Mastery Flip Chart Set 6 to 8
S93938 Math Skills Flip Chart Set 8
S04685 Weather and Sky Flip Chart Set Pre-K to 1
S01467 Curriculum Mastery Flip Chart Set 7
S04688 All About Me Flip Chart Set Pre-K to 1
S04686 Exploring Matter Flip Chart Set Pre-K to 1
S01468 Curriculum Mastery Flip Chart Set 8
S04687 Pushing, Moving and Pulling Flip Chart Set Pre-K to 1
S04682 All About Plants Flip Chart Set Pre-K to 1
S04689 Scient Readiness Flip Chart Set Pre-K to 1
S04683 All About Animals Flip Chart Set Pre-K to 1
S04684 Our Earth Flip Chart Set Pre-K to 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. S01469 Supplier NewPath Learning™ Supplier No. 346012
Add to Cart
Add to Cart

Mounted on a sturdy easel.

  • 10 double-sided, laminated 12 x 18 in. (30.5 x 45.7cm) charts
  • Side 1 features colorful, graphic overview of product
  • Side 2 serves as write-on/wipe-off activity chart featuring questions, labeling exercises, vocabulary review and more
  • Activity Guide features black-line copy-masters and exercises

Lab safety:
Safety Symbols and Labels; Personal Protective Equipment; Safety in the Lab; Hygiene Practices in the Lab; Chemical Exposure; Chemical Handling; Eye Safety; Chemical Spills; Bloodborne Pathogens Safety; Biological Safety

Specifications

Product Type Curriculum Mastery Flip Chart Set
Includes The six kingdoms of life, Introduction to Animals, Sponges, Cnidarians and Worms, Mollusks, Arthropods and Echinoderms, Fishes, Amphibians and Reptiles, Birds and Mammals, Introduction to Plants, roots, Stems and Leaves, Flowers and Seeds, Microlife - Bacteria, Fungi and Protists
Certifications/Compliance Academic Standard for California, Florida, National, New York, Texas
Grade Level Intermediate - Middle School
Grade 6 to 8
Subject Science
Material Laminated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.