Learn More
United Scientific™ Safety Goggles
SDPSCI_ED
Catalog No. S111036 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Lens Coating:
Uncoated
Catalog No. Lens Coating
S111036 Uncoated
1 options
Catalog No. S111036 Supplier United Scientific™ Supplier No. UNGOGGLEP
Add to Cart
Add to Cart
These flexible safety goggles are manufactured from PVC and are designed to protect against splashes and impact.
  • ANSI Z87.1-2020 Certified.
  • Indirect venting to prevent fogging.
  • Adjustable strap for a comfortable fit.
Specifications
Lens Coating Uncoated
Lens Type ANSI Z87.1-2020 Certified
Shape Goggle Shaped
Lens Tint Transparent
Product Type Goggles
For Use With (Application) Eye Protection
Material PVC
Disposable Reusable
Lens Material PVC
Frame Material PVC
UV Protection None
Frame Type Vented Side Arms