Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Safety-Labeled, Two Color Water Wide-Mouth Wash Bottles
GREENER_CHOICESCI_ED

Catalog No. S66409 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
16 oz.
Capacity (Metric):
500 mL
Color:
Natural
Autoclavable:
Not Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Capacity (Metric) Color Autoclavable
S66409 16 oz. 500 mL Natural Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S66409
Add to Cart
Add to Cart

Polyethylene bottles with polypropylene closures to ensure leakproof dispensing.

  • Easy-to-read printed bottles
  • Wide mouth 53 mm (2 in.) bottle for easy refilling
  • Translucent, LDPE solvent-resistant
  • Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 mm
  • Toluene is red LDPE bottle
  • Polypropylene closure with leakproof design
  • Bottles can be purchased separately or in variety packs that include water, acetone, methanol, isopropanol, toluene and ethanol

Compliance

OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Natural
For Use With (Application) Distilled Water
Material LDPE, Polyethylene
Sterility Non-sterile
Closure Color Blue
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide, Threaded
Shape Cylindrical
Product Type Safety-labeled Wash Bottle
Chemical Label Distilled Water
Closure Material Polypropylene
Closure Size 53 mm
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.