Learn More
Caresfield Safety Push Button Butterfly Blood Collection Set
SDP
Catalog No. 22306382
Change view
Click to view available options
Needle Gauge:
21 G
23 G
25 G
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
22-306-382 21 G
22-306-383 23 G
22-306-384 25 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-306-382 Supplier Caresfield Supplier No. 61343CS
Only null left
Add to Cart
Add to Cart
The Safety Push Button Butterfly Blood Collection come in a set of tubes for facilitated blood collection.
  • Push-button needle-retraction safety mechanism
  • Large, ergonomic butterfly wings
  • Double sheaths
  • 200 per case
Specifications
Product Type Safety Blood Collection Needle
Color Green
Material Plastic, Stainless Steel
Needle Gauge 21 G
Sterility Sterile