Learn More
Safety Red-Spirit™ Thermometers
SCI_ED
Catalog No. p-2371646
Change view
Click to view available options
-10 to +200 deg. C/ Sub
Catalog No.
S41574D -10 to +200 deg. C/ Sub
1 options
This item is not returnable. View return policy
Catalog No. S41574D Supplier *S BRANNAN AND SONS Supplier No. 75/360/0EMD
This item is not returnable. View return policy
Solid-stem glass thermometers with dark scale and white background are easy for students to read and can withstand heavy usage.
  • Unique, anti-roll device reduces accidental breakage
  • Individually wrapped in safety seal packs
  • Models with PTFE coating encapsulate the glass and liquid in the event of breakage
  • Premium-grade thermometers have superior quality black markings
  • Student-grade thermometers, most commonly used in school laboratories, have dark brown markings