Learn More
Accuform™ Safety Sign - ATTENTION FOR THE SAFETY AND HEALTH OF OUR CUSTOMERS (Human And Arrows Symbol)
Shop All AccuformNMC ProductsDescription
- Adhesive Vinyl - Printed matte surface has permanent acrylic adhesive back. Moisture resistant for a variety of light-duty indoor and sheltered outdoor marking needs.
- Aluminum - Provides the strength of metal without rusting. Printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.
- Plastic - Durable with slight flex. Good material choice for indoor and sheltered outdoor environments. The printed surface has clear UV protective overcoat.
Specifications
Specifications
| Material | Adhesive Vinyl |
| Mounting Type | Self-adhesive |
| Languages | English |
| Dimensions (L x W) | 14 x 10 in. |
| Displayed Text | ATTENTION FOR THE SAFETY AND HEALTH OF OUR CUSTOMERS AND EMPLOYEES THIS FACILITY PRACTICES SOCIAL DISTANCING PLEASE MAINTAIN DISTANCE BETWEEN YOURSELF AND OTHERS WE APPRECIATE YOUR COOPERATION |
| Color | Red on White |