Learn More
Caresfield Safety Slide Lock Butterfly Blood Collection Set
SDP
Catalog No. 22306391
Change view
Click to view available options
Needle Gauge:
21 G
23 G
25 G
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
22-306-391 21 G
22-306-392 23 G
22-306-393 25 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-306-391 Supplier Caresfield Supplier No. 61634CS
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Safety Slide Lock Butterfly Blood Collection come in a set to facilitate blood collection. Contains a lock.
  • Slide-lock safety mechanism
  • Large, ergonomic butterfly wings
  • Double sheaths
  • 200 per case
Specifications
Product Type Safety Blood Collection Needle
Color Green
Material Plastic, Stainless Steel
Needle Gauge 21 G
Sterility Sterile