Learn More
Saflufenacil Metabolite M800H11, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 1246768-30-8 |
| Linear Formula | C15H13 ClF4N4O5 S |
| Synonym | N′-[2-Chloro-5-(2,6-dioxo-4-(trifluoromethyl)-3,6-dihydro-1(2 H)-pyrimidinyl)-4-fluorobenzoyl]-N-isopropylsulfamide; 2-Chloro-5-[3,6-dihydro-2,6-dioxo-4-(trifluoromethyl)-1(2 H)-pyrimidinyl]-4-fluoro-N-[[(1-methylethyl)amino]sulfonyl]benzamide |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 472.80 |
| Percent Purity | ≥95.0% (HPLC) |
| Grade | Analytical Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.