Learn More
At Battery Company Inc Saft LS14500 AA 3.6V Primary Lithium Battery
Supplier: At Battery Company Inc LS14500
Saft LS14500 primary lithium cell, is based on the Lithium-Thionyl chloride (Li-SOCl2) chemistry. The LS14500 key features are the concentric electrodes (€œbobbin€) construction that enables maximum loading of active materials and corresponding energy. The LS14500 cell is designed specifically for long-term (3 to 15 years) applications, featuring a few UA based currents and periodic pulses, typically in the 5-150 mA range.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.