Learn More
AdipoGen TPh A (1 mg)
SDP
Supplier:  AdipoGen AGCR13507M001
Encompass_Preferred
Chemical. CAS: 21306-65-0. Formula: C21H21NO3S2. MW: 399.5. Nuclear protein pirin inhibitor. Inhibits the interaction between Bcl3 oncoprotein and the nuclear protein pirin. Melanoma cell migration inhibitor.
Catalog No. 50-205-3884
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.