Learn More
Abcam Sag Smo Agonist, Potent, Selective, Solid, 5 mg
Description
- Potent, selective, and cell-permeable Smoothened agonist
- EC50 = 3 nM
- Binds to the Smo heptahelical bundle to antagonize cyclopamine action
- Active in vitro and in vivo
- Purity: >98%
- Molecular weight: 490.1 Da
- CAS Number: 364590-63-6
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Solid |
| Content And Storage | Store at -20°C. |
| Target Molecule | Smoothened |
| Shipping Condition | Ambient |
| Formulation | 100% SAG, Smo Agonist |
| Quantity | 5 mg |
| For Use With (Application) | Smoothened agonist |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.