Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saikosaponin D | 20874-52-6 | MFCD09028095 | >=90.0% | 780.98 g/mol | C42H68O13 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN025010MG

Encompass_Preferred

Saikosaponin D is a triterpene saponin isolated from Bupleurum used in research for its anti-inflammatory, antibacterial, antitumor, and anti-allergic activities. It has been reported to inhibit selectin-mediated binding and downstream signaling pathways including STAT3 and NF-κB, and to activate estrogen receptor-β.

  • Triterpene saponin with multiple reported bioactivities including anti-inflammatory and antitumor effects.
  • Suitable for in vitro and preclinical research at small mg scale.
  • Molecular formula C42H68O13; molecular weight 780.98 g/mol.
  • CAS number 20874-52-6 for unambiguous identification.
  • Purity reported ≥90% by HPLC in vendor reference.

Catalog No. 50-002-95884


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.