Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Salbutamol hemisulfate
GREENER_CHOICESDP

Catalog No. 0634250
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-342-50 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-342-50 Supplier Tocris Bioscience™ Supplier No. 0634/250

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

β2-adrenoceptor agonist

Salbutamol hemisulfate is a selective β2-adrenergic agonist with 29-fold higher selectivity for β2 over β1 receptors. Salbutamol elevates fibroblast growth factor 20 (FGF20) in nigrostriatal neurons, and is neuroprotective in a mouse model of Parkinson's disease.

Chemical Identifiers

CAS 51022-70-9
Molecular Formula C13H23NO7S

Specifications

Quantity 50 mg
Formula Weight 576.7
Purity >99%
Molecular Weight (g/mol) 337.387
Chemical Name or Material Salbutamol hemisulfate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.